Skip to Content
Merck
All Photos(1)

Key Documents

63406

Sigma-Aldrich

Methyl malonyl chloride

purum, ≥97.0% (AT)

Synonym(s):

Methyl chloroformylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCOCH2COCl
CAS Number:
Molecular Weight:
136.53
Beilstein:
1754078
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (AT)

refractive index

n20/D 1.432 (lit.)
n20/D 1.432

bp

57-59 °C/12 mmHg (lit.)

density

1.273 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

storage temp.

2-8°C

SMILES string

COC(=O)CC(Cl)=O

InChI

1S/C4H5ClO3/c1-8-4(7)2-3(5)6/h2H2,1H3

InChI key

UTBCRHAMJFMIIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl malonyl chloride can be used in the synthesis of methyl 7-hydroxy[1,3]thiazolo[5,4-b]pyridin-5(4H)-one-6-carboxylates and 6-carboxamides, which can act like HIV integrase strand transfer inhibitors. It can also be used in the synthesis of Strychnos alkaloids like (-)-leuconicine A and (-)-leuconicine B.

Caution

may discolor to yellow on storage

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Synthesis and HIV-integrase strand transfer inhibition activity of 7-hydroxy [1, 3] thiazolo [5, 4-b]pyridin-5 (4H)-ones"
Boros.EE, et al.
Bioorganic & Medicinal Chemistry, 16(21), 5668-5672 (2006)
"Total synthesis of (-)-leuconicine A and B"
Sirasani G and Andrade.BR
Organic Letters, 13(17), 4736-4737 (2011)
Elif Okutan et al.
Journal of fluorescence, 26(6), 2333-2343 (2016-11-01)
Colorimetric fluorescent chemosensors 4 and 5 based on mono- and di- styryl borondipyrromethenes (BODIPY) linked methyl malonyl were designed for detection of hemoglobin (HgB). Their sensing behavior toward various analytes (Br

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service