Skip to Content
Merck
All Photos(1)

Documents

151386

Sigma-Aldrich

Fumaryl chloride

95%

Synonym(s):

(2E)-2-Butenedioyl dichloride, Fumaric acid chloride, Fumaric acid dichloride, Fumaric dichloride, Fumaroyl dichloride, Fumaryl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCH=CHCOCl
CAS Number:
Molecular Weight:
152.96
Beilstein:
1721342
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

161-164 °C (lit.)

density

1.415 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)\C=C\C(Cl)=O

InChI

1S/C4H2Cl2O2/c5-3(7)1-2-4(6)8/h1-2H/b2-1+

InChI key

ZLYYJUJDFKGVKB-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photodissociation dynamics of fumaryl chloride was studied.

Application

Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sepideh Koosehgol et al.
Materials science & engineering. C, Materials for biological applications, 79, 66-75 (2017-06-21)
In the present study, polyethylene glycol fumarate (PEGF) was synthesized as a component of blend solutions via polycondensation polymerization and characterized by different tests in order to determine its functional groups and its physical properties included melting and crystallization temperature
S J Peter et al.
Journal of biomaterials science. Polymer edition, 10(3), 363-373 (1999-04-03)
To synthesize high molecular weight poly(propylene fumarate) (PPF), fumaryl chloride and propylene glycol were reacted in the presence of potassium carbonate, which serves as a proton scavenger. Transesterification of the resulting low molecular weight oligomer led to a polymer with
Elham Shamirzaei Jeshvaghani et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 106(6), 2371-2383 (2017-11-24)
With regard to flexibility and strength properties requirements of soft biological tissue, elastomeric materials could be more beneficial in soft tissue engineering applications. The present work investigates the use of an elastic polymer, (polycaprolactone fumarate [PCLF]), for fabricating an electrospun
Irrum Mushtaq et al.
Materials science & engineering. C, Materials for biological applications, 108, 110456-110456 (2020-01-12)
Under different pathological conditions, high levels of reactive oxygen species (ROS) cause substantial damage to multiple organs. To counter these ROS levels in multiple organs, we have engineered highly potent novel terpolymers. We found that combination of FDA-approved polyethylene glycol
Monali Kawade et al.
The journal of physical chemistry. A, 115(9), 1538-1546 (2011-02-17)
The photodissociation dynamics of fumaryl chloride (ClCO-CH═CH-COCl) has been studied in a supersonic molecular beam around 235 nm using resonance enhanced multiphoton ionization (REMPI) time-of-flight (TOF) technique by detecting the nascent state of the primary chlorine atom. A single laser

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service