Skip to Content
Merck
All Photos(2)

Key Documents

53363

Supelco

Resorcinol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

product line

TraceCERT®

autoignition temp.

1126 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Resorcinol, a dihydric phenol or a phenyl derivative, is a versatile chemical compound, utilized extensively in a variety of applications including cosmetics, pharmaceuticals, agricultural chemicals, rubber/wood industries, flame retardants, manufacturing of specialty chemicals, antioxidants, etc.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: allium zingiber

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio B Fuertes et al.
Chemical communications (Cambridge, England), 48(49), 6124-6126 (2012-05-15)
Core@shell spheres made up of a thin layer of resorcinol-formaldehyde enveloping a silica core were prepared by means of a one-step method under Stöber conditions. These spheres are used as a platform for the synthesis of carbon or polymeric capsules
Fotini Karavassili et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 12), 1632-1641 (2012-11-16)
The effects of the ligands phenol and resorcinol on the crystallization of human insulin have been investigated as a function of pH. Powder diffraction data were used to characterize several distinct polymorphic forms. A previously unknown polymorph with monoclinic symmetry
Phosphate-functionalized carbon monoliths from deep eutectic solvents and their use as monolithic electrodes in supercapacitors.
Daniel Carriazo et al.
ChemSusChem, 5(8), 1405-1409 (2012-07-06)
Michael R L Stratford et al.
Bioorganic & medicinal chemistry, 21(5), 1166-1173 (2013-01-29)
The inactivation of tyrosinase by resorcinol (1,3-dihydroxybenzene) and seventeen simple derivatives has been investigated using combined spectrophotometry and oximetry together with hplc/ms examination of the oxidation products. The results are consistent with a Quintox mechanism, analogous to that proposed for
Alastair B Ross
Journal of agricultural and food chemistry, 60(36), 8954-8962 (2012-08-14)
Alkylresorcinols are phenolic lipids, with homologues ranging from C17 to C25, found in high concentrations in whole grain wheat and rye, lower concentrations in barley, and negligible concentrations in refined wheat flour. The analysis of alkylresorcinols is of importance due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service