
Resorcinol

Synonym(s):
Resorcinol, 1,3-Benzenediol
Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing