Skip to Content
Merck
All Photos(1)

Key Documents

74347

Supelco

Hydroquinone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

product line

TraceCERT®

autoignition temp.

930 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

285 °C (lit.)

mp

172-175 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaqi Hu et al.
Biosensors & bioelectronics, 47, 45-49 (2013-04-03)
A novel light "on-off" photoelectrochemical sensing for the sensitive determination of hydroquinone was developed based on the porphyrin-functionalized Au nanoparticles on graphene (porphyrin/AuNPs/graphene). Gold nanoparticles (AuNPs) were firstly modified onto graphene sheets using NaBH4 as reductant and the porphyrin/AuNPs/graphene nanocomposites
Amanda J Reig et al.
Nature chemistry, 4(11), 900-906 (2012-10-24)
De novo proteins provide a unique opportunity to investigate the structure-function relationships of metalloproteins in a minimal, well-defined and controlled scaffold. Here, we describe the rational programming of function in a de novo designed di-iron carboxylate protein from the Due
P Carbonnelle et al.
Toxicology and applied pharmacology, 132(2), 220-226 (1995-06-01)
Decreased interleukin-1 (IL-1) production by mononuclear phagocytes has been shown to contribute to benzene myelotoxicity in animals. The study presented here was designed to examine the relevance of this mechanism in humans. Fresh human blood monocytes were exposed to 0.001-10
Douglas McGregor
Critical reviews in toxicology, 37(10), 887-914 (2007-11-21)
The toxicology of hydroquinone has been reviewed on a number of previous occasions. This review targets its potential for carcinogenicity and possible modes of carcinogenic action. The evaluation made by IARC (1999) of its carcinogenic risk to humans was that
A P DeCaprio
Critical reviews in toxicology, 29(3), 283-330 (1999-06-24)
Hydroquinone (HQ) is a high-volume commodity chemical used as a reducing agent, antioxidant, polymerization inhibitor, and chemical intermediate. It is also used in over-the-counter (OTC) drugs as an ingredient in skin lighteners and is a natural ingredient in many plant-derived

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service