Skip to Content
Merck
All Photos(1)

Key Documents

442388

Supelco

3-Methylcholanthrene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16
CAS Number:
Molecular Weight:
268.35
Beilstein:
1913890
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

280 °C/80 mmHg (lit.)

mp

178-180 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc2cc3c(ccc4ccccc34)c5CCc1c25

InChI

1S/C21H16/c1-13-6-7-15-12-20-17-5-3-2-4-14(17)8-9-18(20)19-11-10-16(13)21(15)19/h2-9,12H,10-11H2,1H3

InChI key

PPQNQXQZIWHJRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methylcholanthrene, a potent polycyclic aryl hydrocarbon receptor agonist, which is a carcinogen that can be obtained as a byproduct of combustion of various organic substances like coal, garbage, gas and oil. It suppresses the proliferation of human umbilical vascular endothelial cells.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3?Methylcholanthrene/Aryl?Hydrocarbon Receptor?Mediated Hypertension Through eNOS Inactivation
Chang C-C, et al.
Journal of Cellular Physiology, 232, 1020-1029 (2017)
Mechanism of p53-dependent apoptosis induced by 3-methylcholanthrene involvement of p53 phosphorylation and p38 MAPK
Kwon YW, et al.
The Journal of Biological Chemistry, 277, 1837-1844 (2002)
Molecular mechanisms of p21 and p27 induction by 3?methylcholanthrene, an aryl?hydrocarbon receptor agonist, involved in antiproliferation of human umbilical vascular endothelial cells
Pang P-H, et al.
Journal of Cellular Physiology, 215, 161-171 (2008)
Attenuation of polycyclic aromatic hydrocarbon (PAH)-mediated pulmonary carcinogenesis in mice by omega-3-fatty acids
Moorthy B, et al.
Faseb Journal, 31, 1067-1064 (2017)
Michele W L Teng et al.
Cancer research, 72(16), 3987-3996 (2012-08-08)
Cancer immunoediting, the process by which the immune system controls tumor growth and shapes tumor immunogenicity, consists of 3 stages: elimination, equilibrium, and escape. The molecular mechanisms that underlie the equilibrium phase, during which the immune system maintains tumor dormancy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service