About This Item
Recommended Products
Assay
99%
mp
151-154 °C (lit.)
solubility
methanol: 25 mg/mL, clear, colorless to yellow
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[I-].Cc1cc[n+](C)cc1
InChI
1S/C7H10N.HI/c1-7-3-5-8(2)6-4-7;/h3-6H,1-2H3;1H/q+1;/p-1
InChI key
YOBTXORLVXZWSR-UHFFFAOYSA-M
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service