Skip to Content
Merck
All Photos(1)

Key Documents

144088

Sigma-Aldrich

4-Hydroxybenzaldehyde

98%

Synonym(s):

4-Formylphenol, 4-Hydroxybenzaldehyde, Parahydroxybenzaldehyde, p-Formylphenol, p-Hydroxybenzaldehyde, p-Oxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CHO
CAS Number:
Molecular Weight:
122.12
Beilstein:
471352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

solid

mp

112-116 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(O)cc1

InChI

1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H

InChI key

RGHHSNMVTDWUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Hydroxybenzaldehyde can be used as a reactant to synthesize:
  • Schiff Bases with optical properties through its reaction with anilines.[1]
  • Vanillin via bromination followed by copper-catalyzed coupling reaction with sodium methoxide.[2]
  • Oligo-4-hydroxybenzaldehyde by an oxidative polycondensation reaction with hydrogen peroxide (H2O2) in an alkaline medium.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

345.2 °F

Flash Point(C)

174 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanillin synthesis from 4-hydroxybenzaldehyde
Taber Douglass F, et al.
Journal of Chemical Education, 84(7), 1158-1158 (2007)
Stereoselective synthesis of (-)-centrolobine
Lee Eun, et al.
Bulletin of the Korean Chemical Society,, 25(11), 1609-1610 (2004)
The synthesis, characterization and thermal stability of oligo-4-hydroxybenzaldehyde
Mart H, et al.
Polymer Degradation and Stability, 83(3), 395-398 (2004)
Nadja Schultz-Jensen et al.
Applied biochemistry and biotechnology, 165(3-4), 1010-1023 (2011-07-06)
The potential of wheat straw for ethanol production after pretreatment with O(3) generated in a plasma at atmospheric pressure and room temperature followed by fermentation was investigated. We found that cellulose and hemicellulose remained unaltered after ozonisation and a subsequent
Hypomethylating agents (HMAs) effect on myelodysplastic/myeloproliferative neoplasm unclassifiable (MDS/MPN-U): single institution experience.
Aref Al-Kali et al.
Leukemia & lymphoma, 1-3 (2018-02-22)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service