Skip to Content
Merck
All Photos(2)

Key Documents

1.09065

Supelco

Perchloric acid solution

c(HClO₄) = 0.1 mol/l (0.1 N), in anhydrous acetic acid, reag. Ph. Eur., USP, ready-to-use volumetric solution for titration in non-aqueous media, Titripur®

Synonym(s):

PCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HClO4
CAS Number:
Molecular Weight:
100.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

Product Name

Perchloric acid, in anhydrous acetic acid, for titrations in non-aqueous media c(HClO4) = 0.1 mol/l (0.1 N), Titripur®, reag. Ph. Eur., reag. USP

Agency

reag. Ph. Eur.
reag. USP

Quality Level

product line

Titripur®

form

liquid

autoignition temp.

485 °C

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

0.1 (20 °C in H2O)

transition temp

flash point 40 °C

density

1.06 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

OCl(=O)(=O)=O

InChI

1S/ClHO4/c2-1(3,4)5/h(H,2,3,4,5)

InChI key

VLTRZXGMWDSKGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Determination of phosphorus in commercially available milk: Utilizing ion chromatography with perchloric acid deproteinization, this application provides a reliable method for analyzing phosphorus content in dairy products, essential for quality control in the food industry (Yoshikawa et al., 2024).

  • Versatile ferrous oxidation-xylenol orange assay for lipoxygenase activity: This high-throughput screening technique leverages the reactivity of perchloric acid in enzymatic assays, enhancing the detection of lipoxygenase activity crucial for biotechnological applications (Chrisnasari et al., 2024).

  • Nanoporous gold with microporous structure: Perchloric acid is used in the electrochemical dealloying process to create nanoporous gold structures, which are significant for their applications in catalysis and sensor technology (Li et al., 2024).

  • Stability indicating reversed-phase-high-performance liquid chromatography method: This method uses perchloric acid for the analysis of pharmaceutical compounds, offering enhanced stability and reliability for drug formulation and safety checks (Chintala, 2024).

  • Development of innovative methodology for determination of 6-thioguanine: By utilizing perchloric acid, this HPLC-PDA-based technique allows for precise medical diagnostics, facilitating better disease management through accurate drug monitoring (Noga et al., 2023).

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.0995 - 0.1005 mol/L
Measurement uncertainty ± 0.0003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 84 potassium hydrogen phthalate) by means of volumetric standard potassium hydrogen phthalate (article number 1.02400), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service