Skip to Content
Merck
All Photos(1)

Documents

1.01408

Supelco

Crystal violet (C.I. 42555)

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Crystal violet (C.I. 42555), Hexamethylenepararosaniline chloride, Gentian violet (C.I. 42555), Hexamethyl-p-rosanilinium, Methyl violet 10 B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30ClN3
CAS Number:
Molecular Weight:
407.98
MDL number:
UNSPSC Code:
41116130
EC Index Number:
208-953-6
NACRES:
NA.21

product name

Crystal violet (C.I. 42555), indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

potency

420 mg/kg LD50, oral (Rat)

composition

dye content, ≥90% spectrophotometric assay (calc. on dried substance)

impurities

<0.10% Michler′s ketone

loss

≤7.5% loss on drying, 110°C, 4 h

pH

2.5-3.5 (20 °C, 10 g/L in H2O)

mp

189-194 °C

solubility

10 g/L

density

1.19 g/cm3 at 20 °C

bulk density

220‑400 kg/m3

λmax

589-594 nm in water

absorbance ratio

0.98-1.20 (Ratio, E (lambdamax - 15 nm)/E (lambdamax +  15 nm))

storage temp.

2-30°C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Analysis Note

Dye content (spectrophotometrically; calc. on dried substance): ≥ 90.0%
Identity (UV/VIS-Spectrum): passes test
Absorbance characteristics (Ratio,E (λmax - 15 nm)/E (λmax +  15 nm)): 0.98 - 1.20
Absorption maximum λmax. (water): 589 - 594 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.002 g/l; water): 2000 - 2450
TLC-Test: passes test
Michler`s ketone: <0.10%
Loss on drying (110 °C, 4 h): ≤ 7.5%
Suitability as indicator (ACS): passes test
Suitability as indicator (Reag. Ph. Eur.): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service