Skip to Content
Merck
All Photos(2)

Documents

32483

Supelco

Voriconazole

VETRANAL®, analytical standard

Synonym(s):

2R,3S-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, UK-109496

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3N5O
CAS Number:
Molecular Weight:
349.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@H](c1ncncc1F)[C@](O)(Cn2cncn2)c3ccc(F)cc3F

InChI

1S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25H,6H2,1H3/t10-,16+/m0/s1

InChI key

BCEHBSKCWLPMDN-MGPLVRAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Voriconazole is a second-generation triazole, and is derived from fluconazole. It has a broad and enhanced antifungal spectrum as compared to other older triazoles. It is mostly well tolerated and is available in both oral and intravenous formulations.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

It has been used as one of the constituent of YPD (1% yeast extract, 2% peptone, 2% glucose) liquid media.
Voriconazole is an antifungal used to treat serious fungal infections. Voriconazole inhibits ergosterol synthesis by inhibiting CYP450-dependent 14-α sterol demethylase resulting in a depletion of ergosterol in fungal cell membranes.
Voriconazole is an antifungal used to treat serious fungal infections. Voriconazole inhibits ergosterol synthesis by inhibiting CYP450-dependent 14-a sterol demethylase resulting in a depletion of ergosterol in fungal cell membranes.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Repr. 2 - STOT RE 1 - STOT RE 2 Oral - STOT SE 2 Oral

Target Organs

Eyes, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convergent evolution of calcineurin pathway roles in thermotolerance and virulence in Candida glabrata.
Chen, Ying-Lien, et al.
G3 (Bethesda, Md.), 2 (6), 675- 691 (2012)
Voriconazole: a new triazole antifungal agent.
Saravolatz, Louis D., Leonard B. Johnson, and Carol A. Kauffman.
Clinical Infectious Diseases, 36 (5), 630-637 (2003)
Voriconazole versus amphotericin B for primary therapy of invasive aspergillosis.
Herbrecht, Raoul, et al.
The New England Journal of Medicine, 347 (6), 408-415 (2002)
Kim Vanstraelen et al.
Antimicrobial agents and chemotherapy, 58(11), 6782-6789 (2014-09-04)
Setting the adequate dose for voriconazole is challenging due to its variable pharmacokinetics. We investigated the impact of hypoalbuminemia (<35 g/liter) on voriconazole pharmacokinetics in adult intensive care unit (ICU) patients treated with voriconazole (20 samples in 13 patients) as
Yongxuan Hu et al.
Mycopathologia, 179(1-2), 103-109 (2014-11-05)
Chromoblastomycosis, a chronic fungal infection of skin and subcutaneous tissue caused by dematiaceous fungi, is associated with low cure and high relapse rates. Among all factors affecting clinical outcome, etiological agents have an important position. In southern China, Fonsecaea pedrosoi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service