Skip to Content
Merck
All Photos(1)

Documents

22092

Millipore

Tryptic Soy Broth - Dehydrated Culture Media

NutriSelect® Plus, powder, suitable for microbiology

Synonym(s):

CASO Broth, Soybean Casein digest Broth, TSB, Tryptone Soy Broth, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171607
NACRES:
NA.74

Agency

according to ISO 10273:2017
according to ISO 21871:2006

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein peptone (pancreatic), 17 g/L
dipotassium hydrogen phosphate, 2.5 g/L
glucose, 2.5 g/L
sodium chloride, 5 g/L
soya peptone (papain digest.), 3 g/L

packaging

bottle of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

color

light yellow

final pH

7.3±0.2 (25 °C)

application(s)

bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
pathogen testing
personal care
pharmaceutical
sample preparation
veterinary
water monitoring

microbiology

suitability

nonselective for Bacillus spp.
nonselective for Bacteroides spp.
nonselective for Campylobacter spp.
nonselective for Candida spp.
nonselective for Escherichia coli
nonselective for Neisseria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth is a general-purpose culture medium for the isolation and cultivation of fastidious and non-fastidious microorganisms or the maintenance of stock culture. It is a complex medium suitable for the cultivation and isolation of a wide range of bacteria, yeasts, and molds. The formulation of the TSB medium contains a combination of casein and soy peptones supplying organic nitrogen, particularly amino acids, and long-chained peptides. Sodium chloride maintains the osmotic equilibrium, and the natural sugars from soy peptone are the energy sources.

Application

Tryptic Soy Broth is a medium that supports the luxuriant growth of many fastidious organisms without adding serum. It is used for confirmation of Campylobacter jejuni by motility test. It is suitable for the cultivation of both aerobic and anaerobic bacteria.

Features and Benefits

NutriSelect® Plus are powdered culture media offering
  • Quality control with growth promotion test according to industry standards
  • Quality assurance acc. to ISO 9001
  • Cost-efficient powdered media

Preparation Note

Suspend 30 g of dehydrated media in 1 litre of purified filtered water. Heat with frequent agitation and boil for one minute. Sterilize at 121°C for 15 minutes. Cool to 45-50°C. Mix gently and dispense into sterile Petri dishes or sterile culture tubes.

Other Notes

Capability of selective media to detect heat-injured Shigella flexneri

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.L. Smith et al.
Journal of Food Protection, 53, 141-141 (1990)
Lucia Hrncirova et al.
Folia microbiologica, 64(4), 497-508 (2019-01-19)
The aim of this work was to test the hypothesis that antimicrobial food additives may alter the composition of human gut microbiota by selectively suppressing the growth of susceptible gut microbes. To explore the influence of antimicrobial food additives on
S K Urwyler et al.
Letters in applied microbiology, 62(2), 130-137 (2015-11-20)
Fast microbial identification is becoming increasingly necessary in industry to improve microbial control and reduce biocide consumption. We compared the performances of two systems based on MALDI-TOF MS (VITEK MS and BIOTYPER) and two based on biochemical testing (BIOLOG, VITEK
Diego Alem et al.
World journal of microbiology & biotechnology, 36(8), 120-120 (2020-07-19)
We studied the production and the potential use of a purple-pigment produced by an Antarctic bacterial isolate. This pigment was identified as violacein, a metabolite produced by many bacterial strains and reported that it has antiproliferative activity in many cell
Daniel Hernandez-Patlan et al.
Frontiers in microbiology, 9, 1289-1289 (2018-07-06)
In the present study, in vitro assays were conducted to evaluate the solubility of curcumin (CUR) alone or with polyvinylpyrrolidone (PVP) at different pH, as well as its permeability in Caco-2 cells. Results confirmed that the solid dispersion of CUR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service