Skip to Content
Merck
All Photos(1)

Key Documents

41298

Millipore

Tryptic Soy Broth - Dehydrated Culture Media

NutriSelect® Plus, Vegitone, powder, suitable for microbiology

Synonym(s):

CASO Broth, Vegitone, TSB, Vegitone, Tryptic Soy Broth, Animal-Free, Tryptone Soya Broth, Vegitone Tryptic Soy Broth, Vegitone; Casein Soya Broth, Vegitone; Soybean Casein digest Broth, Vegitone, CASO Broth, Casein Soya Broth, Soybean Casein digest Broth, TSB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171607
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

dextrose, 2.5 g/L
dipotassium phosphate, 2.5 g/L
papaic digest of soyabean meal, 3.0 g/L
sodium chloride, 5.0 g/L
tryptone (vegetable), 17.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
bioburden testing
cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
sterility testing
surface monitoring
veterinary
water monitoring

microbiology

suitability

nonselective for Bacillus spp.
nonselective for Bacteroides spp.
nonselective for Campylobacter spp.
nonselective for Candida spp.
nonselective for Escherichia coli
nonselective for Neisseria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

General description

Vegitone Tryptic Soy Broth is a general purpose medium used for the cultivation of a wide variety of microorganisms. It is recommended for sterility testing of molds and lower bacteria. It contains no material derived from animals.
Vegitone; casein soya broth is a non-selective enrichment media for the cultivation of Streptococci.

Application

Vegitone Tryptic Soy Broth is a general purpose medium used for cultivation of a wide variety of microorganisms. It is recommended for sterility testing of molds and lower bacteria. It contains no material derived from animals.

Preparation Note

Suspend 30 g in 1000 ml distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service