Skip to Content
Merck
All Photos(1)

Key Documents

1.46335

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 250 mL, bottle filling volume 100 mL, closure type, red screw cap with 3 loci, box of 6 bottles

Synonym(s):

CASO, Casein Soybean Digest Broth, SCDB, TSB, Tryptic Soy Broth acc EP+USP, Tryptone Soya Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP 2.6.12
JP 4.05
USP 61

Quality Level

description

For isolation & cultivation of fastidious aerobic bacteria, yeasts, and molds

sterility

sterile

form

liquid

shelf life

12 mo.

feature

closure type red screw cap with 3 loci
ready-to-use

packaging

box of 6 bottles
bottle of

technique(s)

direct inoculation: suitable

bottle capacity

250 mL

bottle filling volume

100 mL

pH

7.3

application(s)

cosmetics
food and beverages
microbiology
pharmaceutical
sterility testing

storage temp.

2-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth is a universal complex medium. It supports the growth of a wide variety of aerobic and facultative anaerobic microorganisms, including fungi by the content of two highly nutritious peptones.

Application

Tryptic Soy Broth is used for the isolation and cultivation of fastidious aerobic bacteria, yeasts and molds. The medium is suitable for sterility testing of non-sterile substances, preparations, and products. It is also used as pre-enrichment broth for the test for specified microorganisms or total viable aerobic count determinations using the MPN method.

Features and Benefits

Our ready to use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service