Skip to Content
Merck
All Photos(3)

Key Documents

146072

Sigma-Aldrich

N,N′-Methylenebis(acrylamide)

99%

Synonym(s):

Bis-acrylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
Flavis number:
7.107
NACRES:
NB.22

vapor density

5.31 (vs air)

Quality Level

Assay

99%

form

powder

mp

>300 °C (lit.)

solubility

water: soluble 20 g/L at 20 °C

storage temp.

2-8°C

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N′-Methylenebis(acrylamide) is a cross linking agent that polymerizes with acrylamide and creates cross links within the polyacrylamide gel. It is capable of creating a network rather than linear chains which helps in maintaining the firmness of gel.

Application

N,N′-Methylenebis(acrylamide) has been used in gel preparation. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
Cross-linking agent for preparation of polyacrylamides.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edwin Makhado et al.
International journal of biological macromolecules, 119, 255-269 (2018-07-23)
In the present study, facile and efficient method was adopted for the synthesis of graft copolymer hydrogel by graft copolymerization of acrylic acid (AA) onto xanthan gum (XG) biopolymer in the presence of N,N'-Methylenebisacrylamide (MBA), and ammonium persulfate (APS) as
Timothy Olding et al.
Journal of medical physics, 36(1), 3-14 (2011-03-25)
This paper explores the combination of cone beam optical computed tomography with an N-isopropylacrylamide (NIPAM)-based polymer gel dosimeter for three-dimensional dose imaging of small field deliveries. Initial investigations indicate that cone beam optical imaging of polymer gels is complicated by
Damla Keskin et al.
Biomacromolecules, 20(1), 243-253 (2018-12-05)
Bacterial infection is a severe problem especially when associated with biomedical applications. This study effectively demonstrates that poly- N-isopropylmethacrylamide based microgel coatings prevent bacterial adhesion. The coating preparation via a spraying approach proved to be simple and both cost and
Roongnapa Suedee et al.
Journal of chromatography. A, 1114(2), 239-249 (2006-03-15)
A temperature sensitive dopamine-imprinted polymer was prepared in 80% aqueous methanol solution by free-radical cross-linking co-polymerisation of methacrylic acid and acrylamide at 60 degrees C in the presence of N,N-methylene-bis-acrylamide as the cross-linker and dopamine hydrochloride as template molecule. The
Ruohong Shi et al.
Small (Weinheim an der Bergstrasse, Germany), 16(37), e2002946-e2002946 (2020-08-11)
Hydrogels with the ability to change shape in response to biochemical stimuli are important for biosensing, smart medicine, drug delivery, and soft robotics. Here, a family of multicomponent DNA polymerization motor gels with different polymer backbones is created, including acrylamide-co-bis-acrylamide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service