Skip to Content
Merck
All Photos(3)

Documents

T9281

Sigma-Aldrich

N,N,N′,N′-Tetramethylethylenediamine

BioReagent, suitable for electrophoresis, ≥99.0%

Synonym(s):

1,2-Bis(dimethylamino)ethane, TEMED, TMEDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1732991
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

product line

BioReagent

Assay

≥99.0%

expl. lim.

9.08 %

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.4179 (lit.)

bp

120-122 °C (lit.)

mp

−55 °C (lit.)

density

0.775 g/mL at 20 °C (lit.)

suitability

suitable for electrophoresis

SMILES string

CN(C)CCN(C)C

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TEMED (N,N,N′,N′-Tetramethylethylenediamine) is molecule which allows rapid polymerization of polyacrylamide gels. In the presence of ammonium persulfate, TEMED is responsible for the formation of free radicals from persulfate, thereby initiating the acrylamide polymerization process. However, excess concentration of TEMED can result in abnormal separation patterns.

Application

TEMED (N,N,N′,N′-Tetramethylethylenediamine) has been used for the preparation of acrylamide hydrogels and sodium dodecyl sulfate-polyacrylamide gels.

Caution

Handle and store under inert gas, air and moisture sensitive.

Analysis Note

Tested as a catalyst for PAGE.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An in vitro Transcription/translation System for Detection of Protein Interaction.
Lin PC et al.
Bio-protocol null
Detecting protein-protein interactions by Far western blotting.
Wu Y et al.
Nature Protocols, 2, 3278-3278 (2007)
Phillips TM
Analytical Techniques in Immunochemistry (1991)
Composite three-dimensional woven scaffolds with interpenetrating network hydrogels to create functional synthetic articular cartilage.
Liao IC et al.
Advances in Functional Materials, 23, 5833-5833 (2013)
Kiyohito Shimura
Electrophoresis, 30(1), 11-28 (2008-12-25)
The methodological advancements in CIEF in tubes and microchips during the period between 2002 and early 2008 are reviewed as a continuation of previous review by the same author (Electrophoresis 2002, 23, 3847-3857). After a brief introduction, the following topics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service