Skip to Content
Merck
All Photos(1)

Key Documents

07326

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium chloride

≥99% (T)

Synonym(s):

N-Butyl-N-methylpyrrolidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20ClN
CAS Number:
Molecular Weight:
177.71
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99% (T)

Quality Level

Assay

≥99% (T)

impurities

≤0.5% water

SMILES string

[Cl-].CCCC[N+]1(C)CCCC1

InChI

1S/C9H20N.ClH/c1-3-4-7-10(2)8-5-6-9-10;/h3-9H2,1-2H3;1H/q+1;/p-1

InChI key

BOOXKGZZTBKJFE-UHFFFAOYSA-M

General description

1-Butyl-1-methylpyrrolidinium chloride (BMIMCl), is an ionic liquid composed of a pyrrolidinium cation and a chloride anion. It is used as a solvent in various chemical reactions, catalysis, and separations due to its ability to dissolve a wide range of organic and inorganic compounds. It is also used as an effective corrosion inhibitor.

Application

1-Butyl-1-methylpyrrolidinium chloride can be used as a solvent in the synthesis of oxychloridoselenites. It can also used as a phase-transfer catalyst in reactions such as oxidation and alkylation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hyun Deog Yoo et al.
Nature communications, 8(1), 339-339 (2017-08-25)
Magnesium rechargeable batteries potentially offer high-energy density, safety, and low cost due to the ability to employ divalent, dendrite-free, and earth-abundant magnesium metal anode. Despite recent progress, further development remains stagnated mainly due to the sluggish scission of magnesium-chloride bond
Mixing thermodynamic properties of 1-butyl-4-methylpyridinium tetrafluoroborate [b4mpy][BF 4] with water and with an alkan-1ol (methanol to pentanol)
Ortega, J., et al.
Physical Chemistry Chemical Physics, 40.7, 1087-1094 (2008)
Experimental Thermodynamic Properties of 1-Butyl-2-methylpyridinium Tetrafluoroborate [b2mpy][BF4] with Water and with Alkan-1-ol and Their Interpretation with the COSMO-RS Methodology
Navas, Ana, et al.
Industrial & Engineering Chemistry Research, 48.5, 2678-2690 (2009)
Influence of the electrode potential and in situ STM scanning conditions on the phase boundary structure of the single crystal Bi (111)| 1-butyl-4-methylpyridinium tetrafluoroborate interface
Anderson, Erik, et al.
Journal of Electroanalytical Chemistry, 709, 46-56 (2013)
Supported room temperature ionic liquid membranes for CO 2/CH 4 separation
Iarikov, D. D., P. Hacarlioglu, and S. T. Oyama
Chemical Engineering Journal, 166.1, 401-406 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service