690597
1-Decyl-3-methylimidazolium chloride
96%
Synonym(s):
[C10MIM][Cl]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H27ClN2
CAS Number:
Molecular Weight:
258.83
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥95.5%
96%
form
liquid
impurities
≤2.0% water
refractive index
n20/D 1.501
SMILES string
[Cl-].CCCCCCCCCCn1cc[n+](C)c1
InChI
1S/C14H27N2.ClH/c1-3-4-5-6-7-8-9-10-11-16-13-12-15(2)14-16;/h12-14H,3-11H2,1-2H3;1H/q+1;/p-1
InChI key
HTZVLLVRJHAJJF-UHFFFAOYSA-M
Related Categories
General description
1-Decyl-3-methylimidazolium chloride is a surface active ionic liquid that can be prepared by reacting 1-methylimidazole with 1-chlorodecane.
Application
1-Decyl-3-methylimidazolium chloride can be used in the preparation of 1-decyl-3-methylimidazolium triflate by reacting with sodium triflate.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Influence of the Hofmeister anions on self-organization of 1-decyl-3-methylimidazolium chloride in aqueous solutions
Luczak, Justyna, et al.
Journal of Colloid and Interface Science, 362.2, 415-422 (2011)
Synthesis and micellar properties of surface-active ionic liquids: 1-alkyl-3-methylimidazolium chlorides.
El Seoud OA, et al.
Journal of Colloid and Interface Science, 3131(1), 296-304 (2007)
Karishma Bakshi et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183103-183103 (2019-11-02)
Here, we report the toxic effects of various imidazolium-based ionic liquids (ILs) with varying hydrocarbon chain lengths, on different human cell lines. Multiple biological assays have shown that the ILs with long hydrocarbon chains have stronger adverse effect especially on
Antibiofilm activities of 1-alkyl-3-methylimidazolium chloride ionic liquids
Carson, Louise, et al.
Green Chemistry, 11.4, 492-497 (2009)
Measurements and Correlation of the (Solid+ Liquid) Equilibria of [1-Decyl-3-methylimidazolium Chloride+ Alcohols (C2? C12)]
Domanska, Urszula, and Ewa Bogel-Lukasik
Industrial & Engineering Chemistry Research, 42.26, 6986-6992 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service