Skip to Content
Merck
All Photos(1)

Documents

38894

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide

≥98.0% (T)

Synonym(s):

BMP-BTI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20F6N2O4S2
CAS Number:
Molecular Weight:
422.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

electrochemical window -3.0 - +2.4 V

Quality Level

Assay

≥98.0% (T)

impurities

≤0.2% water

viscosity

548 stokes

mp

-18 °C

SMILES string

CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C9H20N.C2F6NO4S2/c1-3-4-7-10(2)8-5-6-9-10;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h3-9H2,1-2H3;/q+1;-1

InChI key

HSLXOARVFIWOQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-1-methylpyrrolidinium bis(trifluoromethylsulfonyl)imide is a room temperature ionic liquid (RTIL) with a wide electrochemical window (5.5V) and high viscosity, making it a useful candidate in electrochemical energy applications.

Application

BMP-BTI has been used as a derivatization media for dimethyl sulfate (DMS) with dibenzazepine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ionic Liquids for Energy Storage Applications
Stenger-Smith JD and Irvin JA.
Material Matters, 4(4), 103-103 (2009)
An ionic liquid based strain sensor for large displacement measurement
Keulemans, Grim, Frederik Ceyssens, and Robert Puers
Biomedical Microdevices, 19.1, 1-1 (2017)
Electrochemical reduction of O 2 in 1-butyl-1-methylpyrrolidinium bis (trifluoromethanesulfonyl) imide ionic liquid containing Zn 2+ cations: deposition of non-polar oriented ZnO nanocrystalline films
Azaceta, Eneko, et al.
Physical Chemistry Chemical Physics, 13.29, 13433-13440 (2011)
Electrodeposition of al in 1-butyl-1-methylpyrrolidinium bis (trifluoromethylsulfonyl) amide and 1-ethyl-3-methylimidazolium bis (trifluoromethylsulfonyl) amide ionic liquids: In situ STM and EQCM studies
Moustafa, E. M., et al.
The Journal of Physical Chemistry B, 111.18, 4693-4704 (2007)
Voltammetric Study and Electrodeposition of Zinc in Hydrophobic Room-Temperature Ionic Liquid 1-Butyl-1-methylpyrrolidinium Bis ((trifluoromethyl) sulfonyl) imide ([BMP][TFSI]): A Comparison between Chloride and TFSI Salts of Zinc
Wang, Yu-Sheng, et al.
Journal of the Electrochemical Society, 164.2, D39-D47 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service