About This Item
Recommended Products
grade
analytical standard
for GPC
Quality Level
form
powder or crystals
mol wt
Mn ~236,300
Mp ~276,500
Mw ~409,800
analyte chemical class(es)
oligosaccharides
technique(s)
gel permeation chromatography (GPC): suitable
Mw/Mn
~1.73
application(s)
food and beverages
InChI
1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2
InChI key
FZWBNHMXJMCXLU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Dextran in Drug Delivery: A study in "Journal of Biomedical Materials Research Part A" demonstrated how dextran, a biopolymer, enhances wound healing. It focused on collagen-polyurethane-dextran hydrogels, which inhibit inflammation and promote collagen fibrillogenesis, making dextran crucial for tissue engineering and medical applications (Aguayo-Morales et al., 2024).
- Dextran as a Blood Plasma Expander: Research in "Redox Biology" highlighted the role of dextran in reducing systemic iron overload stress. This study provides insights into how dextran′s properties are beneficial for managing conditions like anemia and other blood volume deficiencies (Zhao et al., 2024).
- Dextran in Cell Culture: An article in ""International Journal of Biological Macromolecules"" explored the use of dextran-based antibacterial hydrogels. These gels are used for fluorescent sensors in bioanalytical applications, showing dextran′s utility in cell culture and microbial monitoring (Bai et al., 2024).
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service