Skip to Content
Merck
All Photos(1)

Key Documents

00892

Supelco

Dextran

analytical standard, for GPC, 80,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

form

powder or crystals

mol wt

Mn ~55,500
Mp ~66,700
Mw ~80,900

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.46

application(s)

food and beverages

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran (mol. wt. 80,000) belongs to the family of neutral polysaccharides, which are recognized as contaminants in sugar processing and other food production industries. These are composed of chains of D-glucose units connected by alpha-(1-6) linkages of varying lengths. They are found as bacterial extracellular polysaccharides and reportedly synthesized from sucrose by beneficial lactic acid bacteria, such as Leuconostoc mesenteroides and Lactobacillus brevis. They are incorporated in bakery products to improve the crumb texture, softness, and loaf volume, as stabilizers and additives in confectionery, flavor extract, icing compositions, beverages including soft drinks, milk, etc.

Application

Dextran has been used as a polymer standard in the quantitative determination of intracellular sodium [Na+] using fluorescence lifetime imaging microscopy (FLIM) with CoroNaGreen dye. It has also been used in combination with concanavalin A as concanavalin A-dextran affinity assay employed in osmotic glucose sensor for the characterization of commercial nanoporous membranes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of cellular [Na+] by fluorescence lifetime imaging with CoroNaGreen
Meyer J, et al.
The Journal of General Physiology, 151(11), 1319-1331 (2019)
Characterization of nanoporous membranes for implementation in an osmotic glucose sensor based on the concanavalin A--dextran affinity assay
Krushinitskaya O, et al.
Journal of Membrane Science , 376(1-2), 153-161 (2011)
Functional polymers based on dextran
Polysaccharide-based Fibers and Composites: Chemical and Engineering Fundamentals and Industrial Applications, 376(1-2), 199-291 (2006)
Dextran?the polysaccharide with versatile uses
Bhavani LA and Nisha J
International Journal of Pharmacy and Biological Sciences, 1(4), 569-573 (2010)
Dmitri Simberg et al.
Biomaterials, 30(23-24), 3926-3933 (2009-04-28)
In order to understand the role of plasma proteins in the rapid liver clearance of dextran-coated superparamagnetic iron oxide (SPIO) in vivo, we analyzed the full repertoire of SPIO-binding blood proteins using novel two-dimensional differential mass spectrometry approach. The identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service