Merck
Carbohydrate Standards
A pile of white sugar cubes neatly stacked against a plain white background. The cubes are uniform in size and shape, suggesting a sense of order and simplicity. The image highlights the granular texture of the sugar cubes.
 Carbohydrates, also known as saccharides, are the macronutrients found as sugars, fibers, and starches in food. They are biomolecules made up of carbon, hydrogen, and oxygen atoms and commonly have a hydrogen and oxygen ratio of 2:1. 
We offer a broad selection of carbohydrate standards ranging from simple to complex carbohydrates for analytical testing, including pharma release testing, pharma method development, and quality control testing of food and beverages. Our standards are available in a variety of quality grades suitable for analysis by chromatographical methods.
Products
Sort by Relevance
Showing 1-20 of 55
Change View
Showing 1-20 of 55
Compare
Product Number
Product Name
Product Description
Pricing
You have viewed 1-20 of 55 results
Monosaccharides
Our offering includes common monosaccharides, such as glucose, fructose, and galactose, as analytical standards or as secondary pharmaceutical standards / certified reference materials.
Disaccharides
Our offering of disaccharide standards includes lactose, sucrose, maltitol, maltose, D-(+)-melibiose, palatinose, D-(+)-turanose, D-(+)-cellobiose, and galactinol dihydrate.
Oligosaccharides
We provide standards for several oligosaccharides such as isomaltotriose, 1-kestose, D-(+)-melezitose, nystose, verbascose, D-(+)-raffinose pentahydrate, beta-cyclodextrin, and acarbose.
Polysaccharides & Starches
Our portfolio includes dextran and pullulan standards for GPC or a standard for starch assay kits.
Carbohydrate Solutions for Ion Chromatography
Our range of carbohydrate standards also includes their certified reference material (CRM) solutions for ion chromatography applications.
  • Manufactured and certified under ISO/IEC 17025 and ISO 17034
  • Provided with a comprehensive certificate showing certified quantitative value determined by mass balance
  • Traceable to the primary standards maintained by National Metrology Institutes (NMI)