Skip to Content
Merck
All Photos(1)

Key Documents

28-5700

Sigma-Aldrich

Sulfamic acid

JIS special grade, ≥99.5%

Synonym(s):

Amidosulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2SO3H
CAS Number:
Molecular Weight:
97.09
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.5%

form

solid

availability

available only in Japan

mp

215-225 °C (dec.) (lit.)

solubility

water: 213 g/L at 20 °C
water: 470 g/L at 80 °C

density

2.151 g/cm3 at 25 °C

SMILES string

NS(O)(=O)=O

InChI

1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cobalt-catalyzed direct arylation and benzylation by C-H/C-O cleavage with sulfamates, carbamates, and phosphates.
Weifeng Song et al.
Angewandte Chemie (International ed. in English), 51(33), 8251-8254 (2012-07-14)
Christopher S Adams et al.
Journal of the American Chemical Society, 134(26), 10807-10810 (2012-06-20)
Nitrogen-containing stereotriads, compounds with three adjacent stereodefined carbons, are commonly found in biologically important molecules. However, the preparation of molecules bearing these motifs can be challenging. Herein, we describe a modular oxidation protocol which converts a substituted allene to a
Laszlo Tarko et al.
Bioorganic & medicinal chemistry, 21(6), 1404-1409 (2012-12-05)
The last version of the PRECLAV algorithm was used to investigate a series of sulfamate/sulfamide carbonic anhydrase (CA, EC 4.2.1.1) inhibitors. PRECLAV allows identification of outliers for lead hopping, significant molecular fragments and similarity computation of a calibration set vs.
Edward J Emmett et al.
Organic & biomolecular chemistry, 10(20), 4007-4014 (2012-03-13)
By using DABCO·(SO(2))(2), DABSO, as a solid bench-stable SO(2)-equivalent, the palladium-catalysed aminosulfonylation of aryl-, alkenyl- and heteroaryl halides has been achieved. N,N-Dialkylhydrazines are employed as the N-nucleophiles and provide N-aminosulfonamides as the products in good to excellent yields. The reactions
Ning Dai et al.
Environmental science & technology, 46(17), 9793-9801 (2012-07-27)
With years of full-scale experience for precombustion CO(2) capture, amine-based technologies are emerging as the prime contender for postcombustion CO(2) capture. However, concerns for postcombustion applications have focused on the possible contamination of air or drinking water supplies downwind by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service