Skip to Content
Merck
All Photos(1)

Key Documents

86040

Supelco

Sulfamic acid

analytical standard (for acidimetry), ACS reagent

Synonym(s):

Aminosulfonic acid, Sulphamic acid, Amidosulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2SO3H
CAS Number:
Molecular Weight:
97.09
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

ACS reagent
analytical standard (for acidimetry)

Quality Level

Agency

complies with DIN 19266

Assay

99.3-100.3% (dried material)

mp

215-225 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

SMILES string

NS(O)(=O)=O

InChI

1S/H3NO3S/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

IIACRCGMVDHOTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfamic acid, a strong inorganic acid, exists in crystalline form. It is stable, non-hygroscopic, odorless, and colorless in dry state, making its commercial packaging and handling convenient.

Application

It can be used as an acidimetric standard for analytical applications.
Sulfamic acid also finds its use as a primary standard in non-aqueous visual, conductometric, and potentiometric titrations.

Features and Benefits

  • Available in a secure glass bottle to ensure its stability for the entire shelf life until opened.
  • High-quality offering accurate titer determinations
  • Accompanied by a detailed certificate of analysis (CoA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derek J Langeslay et al.
Analytical chemistry, 83(20), 8006-8010 (2011-09-15)
Sulfamate (NHSO(3)(-)) groups are critically important structural elements of the glycosaminoglycans heparin and heparan sulfate (HS). Experimental conditions are presented for detection of the sulfamate (1)H NMR resonances in aqueous solution. NMR spectra reported for N-sulfoglucosamine (GlcNS) and the synthetic
Toolika Agrawal et al.
Organic letters, 15(1), 96-99 (2012-12-19)
The iron-catalyzed cross-coupling of aryl sulfamates and tosylates has been achieved with primary and secondary alkyl Grignards. This study of iron-catalyzed cross-coupling reactions also examines the isomerization and β-hydride elimination problems that are associated with the use of isopropyl nucleophiles.
Daniel Can et al.
Angewandte Chemie (International ed. in English), 51(14), 3354-3357 (2012-02-22)
Enhanced receptor selectivity: carbonic anhydrase inhibitors are relevant for both cancer diagnosis and therapy. Combining non-radioactive Re compounds with their radioactive (99m)Tc homologs enables the use of identical molecules for therapy and imaging (theragnostic). The syntheses and in vitro evaluation
Jean-Yves Winum et al.
Bioorganic & medicinal chemistry, 21(6), 1419-1426 (2012-12-04)
Targeting tumour associated carbonic anhydrase (CA, EC 4.2.1.1) isoforms IX and XII is now considered as a pertinent approach for the development of new cancer therapeutics against hypoxic tumours. In the last period, with the help of X-ray crystallography, much
Cobalt-catalyzed direct arylation and benzylation by C-H/C-O cleavage with sulfamates, carbamates, and phosphates.
Weifeng Song et al.
Angewandte Chemie (International ed. in English), 51(33), 8251-8254 (2012-07-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service