Skip to Content
Merck
All Photos(1)

Key Documents

07-6060

Sigma-Aldrich

Distilled water

suitable for HPLC

Synonym(s):

Water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
100 °C (lit.)

grade

HPLC grade

form

liquid

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucio Isa
Chimia, 67(4), 231-235 (2013-08-24)
Since the seminal work of Pickering and Ramsden more than a century ago, adsorption of solid micro- and nanoparticles at the interface between two fluids has been recognized as a means to enormously improve emulsion stability against coalescence. Despite their
Steven Brantley et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 777-790 (2013-07-20)
Infestation of eastern hemlock (Tsuga canadensis (L.) Carr.) with hemlock woolly adelgid (HWA, Adelges tsugae) has caused widespread mortality of this key canopy species throughout much of the southern Appalachian Mountains in the past decade. Because eastern hemlock is heavily
Anthony W D'Amato et al.
Ecological applications : a publication of the Ecological Society of America, 23(8), 1735-1742 (2014-02-22)
Reducing tree densities through silvicultural thinning has been widely advocated as a strategy for enhancing resistance and resilience to drought, yet few empirical evaluations of this approach exist. We examined detailed dendrochronological data from a long-term (> 50 years) replicated
Jesse K Kreye et al.
Ecological applications : a publication of the Ecological Society of America, 23(8), 1976-1986 (2014-02-22)
Long-term fire exclusion has altered ecological function in many forested ecosystems in North America. The invasion of fire-sensitive tree species into formerly pyrogenic upland forests in the southeastern United States has resulted in dramatic shifts in surface fuels that have
Liming Zhou et al.
Nature, 509(7498), 86-90 (2014-04-25)
Tropical forests are global epicentres of biodiversity and important modulators of climate change, and are mainly constrained by rainfall patterns. The severe short-term droughts that occurred recently in Amazonia have drawn attention to the vulnerability of tropical forests to climatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service