Skip to Content
Merck
All Photos(1)

Key Documents

01-0650

Sigma-Aldrich

Acetonitrile

≥99.8%, suitable for HPLC

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
41116023
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

grade

HPLC grade

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Preparation and characterization of stationary phase gradients on C8 liquid chromatography columns.: This study explores the development and analysis of stationary phase gradients on C8 columns, crucial for optimizing chromatographic separations in analytical chemistry (Cecil et al., 2024).

  • Metal-cation-induced shifts in thiolate redox and reduced sulfur speciation.: Investigates the impact of metal cations on the redox behavior of thiolates, a key area in understanding redox processes in various chemical systems (Seo et al., 2024).

  • A Phosphine-Oxide Cobalt(II) Complex and Its Catalytic Activity Studies toward Alcohol Dehydrogenation Triggered Direct Synthesis of Imines and Quinolines.: Describes the catalytic role of a cobalt(II) phosphine-oxide complex in synthesizing imines and quinolines from alcohols, a significant contribution to synthetic organic chemistry (Khatua et al., 2024).

  • [Simultaneous determination of 31 banned veterinary drugs during egg-laying period in poultry eggs by ultra performance liquid chromatography-tandem mass spectrometry].: This Chinese study presents a method for detecting multiple veterinary drugs in eggs, highlighting advancements in food safety and analytical methodologies (Zhu et al., 2024).

  • Probing Isolated Water Molecules in Aqueous Acetonitrile Solutions Using Oxygen K-Edge X-ray Absorption Spectroscopy.: Focuses on the molecular dynamics of water in acetonitrile solutions, providing insights that are crucial for understanding solvent effects in various chemical environments (Nagasaka, 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.0 °F

Flash Point(C)

12.8 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic
Fang Xu et al.
The Plant cell, 27(5), 1547-1562 (2015-05-15)
Nod-like receptors (NLRs) serve as immune receptors in plants and animals. The stability of NLRs is tightly regulated, though its mechanism is not well understood. Here, we show the crucial impact of N-terminal acetylation on the turnover of one plant
Kangmin Chon et al.
Water research, 81, 388-397 (2015-07-05)
In this study, the changes in UV absorbance at 254 nm (UVA254) and electron donating capacity (EDC) were investigated as surrogate indicators for assessing removal of micropollutants and bromate formation during ozonation of wastewater effluents. To measure the EDC, a novel
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service