Skip to Content
Merck
All Photos(1)

Key Documents

07-4770

Sigma-Aldrich

Dimethyl phthalate

SAJ special grade, ≥99.0%

Synonym(s):

DMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
Beilstein:
1911460
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

liquid

autoignition temp.

1033 °F

availability

available only in Japan

refractive index

n20/D 1.515 (lit.)

bp

282 °C (lit.)

mp

2 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccccc1C(=O)OC

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenxin Wang et al.
Environmental monitoring and assessment, 184(11), 6789-6798 (2011-12-08)
Atmospheric trace phthalic esters (PAEs) carried by total suspended particulates were systematically investigated. A total of 450 air samples were collected at six typical locations and three specific micro-environmental sites (a stadium with a new plastic track, new cars, and
L J Xu et al.
Water research, 47(6), 1996-2004 (2013-02-12)
The merits of the combined process of high-frequency ultrasound (US) and catalyst-free ultraviolet irradiation (UV) have been evaluated in this study by investigating the sonophotolytic degradation of dimethyl phthalate (DMP). A 400 kHz ultrasonic system and a photolytic system at
Yiling Chen et al.
Journal of hazardous materials, 235-236, 92-100 (2012-08-14)
In this study, we demonstrate that the decomposition of dimethyl phthalate under microwave irradiation could be greatly enhanced over Fe@Fe(2)O(3) nanowires supported on activated carbon (Fe@Fe(2)O(3)/AC). The great enhanced decomposition of dimethyl phthalate could be attributed to a unique microwave
L J Xu et al.
Ultrasonics sonochemistry, 20(3), 892-899 (2012-12-05)
A comprehensive study of the sonochemical degradation of dimethyl phthalate (DMP) was carried out using high-frequency ultrasonic processes. The effects of various operating parameters were investigated, including ultrasonic frequency, power density, initial DMP concentration, solution pH and the presence of
Zhujian Huang et al.
Journal of hazardous materials, 246-247, 70-78 (2013-01-05)
The organic layered double hydroxides (LHDs)/TiO(2) composites with various mass ratios were prepared by the reconstruction of mixed metal oxides to photodegrade dimethyl phthalate (DMP). The physicochemical properties of the obtained products were analyzed by X-ray diffraction (XRD) spectra, X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service