Skip to Content
Merck
Dimethyl phthalate SAJ special grade, ≥99.0%

Dimethyl phthalate

Dimethyl phthalate SAJ special grade, ≥99.0%
Synonym(s):
DMP, Dimethyl phthalate
Linear Formula:
C6H4-1,2-(CO2CH3)2
CAS Number:
Molecular Weight:
194.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing