Skip to Content
Merck
All Photos(1)

Key Documents

05-7200

Sigma-Aldrich

Cyclohexanone

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

availability

available only in Japan

refractive index

n20/D 1.450 (lit.)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of fragment screening and merging to the discovery of inhibitors of the Mycobacterium tuberculosis cytochrome P450 CYP121.
Sean A Hudson et al.
Angewandte Chemie (International ed. in English), 51(37), 9311-9316 (2012-08-15)
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2629-2637 (2012-09-07)
Iron (and to a lesser extent manganese) in the wall of a 316 stainless steel (SS) reactor is responsible for the hydrogenation of cyclohexanone to cyclohexanol when using an aqueous formic acid solution under high temperature and pressure water (HTPW)
Alakananda Hajra et al.
Organic letters, 14(21), 5488-5491 (2012-10-18)
Dehydrogenative aromatization of cyclohexanone imines to arylamines has been achieved using a palladium catalyst under aerobic conditions. The reaction is applicable to a variety of imines that are either preformed or generated in situ from cyclohexanone derivatives and aryl or
Won-Heong Lee et al.
Applied microbiology and biotechnology, 97(4), 1561-1569 (2012-10-12)
Sufficient supply of NADPH is one of the most important factors affecting the productivity of biotransformation processes. In this study, construction of an efficient NADPH-regenerating system was attempted using direct phosphorylation of NADH by NADH kinase (Pos5p) from Saccharomyces cerevisiae
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2638-2643 (2012-09-07)
The processing of renewable feedstocks to platform chemicals and, to a lesser degree, fuels is a key part of sustainable development. In particular, the combination of lignocellulosic biomass with hydrothermal upgrading (HTU), using high temperature and pressure water (HTPW), is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service