Merck
  • Effects of NADH kinase on NADPH-dependent biotransformation processes in Escherichia coli.
Effects of NADH kinase on NADPH-dependent biotransformation processes in Escherichia coli.
Applied microbiology and biotechnology (2012-10-12)
Won-Heong Lee, Jin-Woo Kim, Eun-Hee Park, Nam Soo Han, Myoung-Dong Kim, Jin-Ho Seo
ABSTRACT
Sufficient supply of NADPH is one of the most important factors affecting the productivity of biotransformation processes. In this study, construction of an efficient NADPH-regenerating system was attempted using direct phosphorylation of NADH by NADH kinase (Pos5p) from Saccharomyces cerevisiae for producing guanosine diphosphate (GDP)-L-fucose and ε-caprolactone in recombinant Escherichia coli. Expression of Pos5p in a fed-batch culture of recombinant E. coli producing GDP-L-fucose resulted in a maximum GDP-L-fucose concentration of 291.5 mg/l, which corresponded to a 51 % enhancement compared with the control strain. In a fed-batch Baeyer-Villiger (BV) oxidation of cyclohexanone using recombinant E. coli expressing Pos5p, a maximum ε-caprolactone concentration of 21.6 g/l was obtained, which corresponded to a 96 % enhancement compared with the control strain. Such an increase might be due to the enhanced availability of NADPH in recombinant E. coli expressing Pos5p. These results suggested that efficient regeneration of NADPH was possible by functional expression of Pos5p in recombinant E. coli, which can be applied to other NADPH-dependent biotransformation processes in E. coli.
Sigma-Aldrich
Cyclohexanone, SAJ first grade, ≥98.0%
Sigma-Aldrich
Cyclohexanone, JIS special grade, ≥99.0%
Supelco
Cyclohexanone, Selectophore, ≥99.5%
Sigma-Aldrich
Cyclohexanone, ACS reagent, ≥99.0%
Supelco
Cyclohexanone, analytical standard
Sigma-Aldrich
Cyclohexanone, 99.8%
Sigma-Aldrich
Cyclohexanone, ReagentPlus®, 99.8%
Sigma-Aldrich
Cyclohexanone, puriss. p.a., ≥99.5% (GC)