Skip to Content
Merck
All Photos(1)

Key Documents

01-4750

Sigma-Aldrich

Ammonium iodide

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4I
CAS Number:
Molecular Weight:
144.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

vapor pressure

1 mmHg ( 210.9 °C)

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

4.5-6.5 (25 °C, 50 g/L)

SMILES string

N.I

InChI

1S/HI.H3N/h1H;1H3

InChI key

UKFWSNCTAHXBQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aline C A Silva et al.
Analytical and bioanalytical chemistry, 407(14), 4091-4099 (2015-04-08)
This work evaluates the application of a thermodynamic model to comprehensive two-dimensional gas chromatography (GC × GC) coupled with time-of-flight mass spectrometry for anabolic agent investigation. Doping control deals with hundreds of drugs that are prohibited in sports. Drug discovery
Junseok Lee et al.
Chemosphere, 142, 72-76 (2015-06-29)
A novel approach using wavelength dispersive X-ray fluorescence (WDXRF) spectrometry combined with an activated carbon (AC) disk was developed for the determination of total iodine concentrations in water samples. Dissolved iodine species (i.e., I(-) and IO3(-)) in water samples were
Ebru Uçaktürk
Journal of analytical methods in chemistry, 2015, 707414-707414 (2015-12-19)
A sensitive and selective gas chromatography-mass spectrometry (GC-MS) method was developed and fully validated for the determination of vildagliptin (VIL) in pharmaceutical formulation. Prior to GC-MS analysis, VIL was efficiently derivatized with MSTFA/NH4I/β-mercaptoethanol at 60°C for 30 min. The obtained O-TMS
M Raro et al.
Journal of mass spectrometry : JMS, 49(6), 509-521 (2014-06-11)
The detection of anabolic androgenic steroids (AAS) is one of the most important topics in doping control analysis. Gas chromatography coupled to (tandem) mass spectrometry (GC-MS(/MS)) with electron ionization and liquid chromatography coupled to tandem mass spectrometry have been traditionally
Eung Ju Kim et al.
PloS one, 9(6), e97841-e97841 (2014-06-06)
Although triglyceride accumulation in the liver causes non-alcoholic fatty liver disease (NAFLD), hypercholesterolemia is also a main cause of NAFLD as well as atherosclerosis. However, NAFLD and atherosclerosis have not been investigated simultaneously in animal models fed a high-cholesterol diet.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service