MAB345A4
Anti-O4 Antibody, clone 81, Alexa Fluor™488 Conjugate
clone 81 (mAB 04), from mouse, ALEXA FLUOR™ 488
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
ALEXA FLUOR™ 488
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
81 (mAB 04), monoclonal
species reactivity
rat
species reactivity (predicted by homology)
human (based on 100% sequence homology), bovine (based on 100% sequence homology), chicken (based on 100% sequence homology), mouse (based on 100% sequence homology)
technique(s)
immunocytochemistry: suitable
isotype
IgM
shipped in
wet ice
target post-translational modification
unmodified
General description
Specificity
Immunogen
Application
Neuroscience
Neurodegenerative Diseases
Quality
Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected O4 in undifferentiated rat oligodendrocyte precursor cells.
Physical form
Storage and Stability
Analysis Note
Undifferentiated rat oligodendrocyte precursor cells
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service