Skip to Content
Merck
All Photos(1)

Key Documents

MAB342

Sigma-Aldrich

Anti-Galactocerebroside Antibody, clone mGalC

clone mGalC, Chemicon®, from mouse

Synonym(s):

Galactoceramide, Galactocerebroside, Galactosylceramide, GalC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

clone

mGalC, monoclonal

species reactivity

mouse, human, rabbit, bovine, chicken, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG3

shipped in

dry ice

target post-translational modification

unmodified

General description

Galactocerebroside (GalC) is a major galactosphingolipid of myelin which plays a role in myelination. GalC is a very useful, specific marker for oligodendroglial lineage.

Specificity

Galactocerebroside (GalC), sulfatide, psychosine and other galactolipids. Cross-reacts with the sulfatide ester of GalC, but to a 16-fold lesser extent. No cross-reactivity with sphingosine, ceramide, mixed ganglioside or glucocerebroside. Binds specifically with oligodendrocytes and Schwann cells.

Immunogen

Synaptic plasma membranes from bovine hippocampus. Ranscht, B. et al. PNAS 79(8):2709-2713 (1982)

Application

Anti-Galactocerebroside Antibody, clone mGalC is an antibody against Galactocerebroside for use in ELISA, IC, & IHC.
Immunohistochemistry:
0.5-10 μg/mL of a previous lot was used on formalin fixed frozen sections of a prevous lot. Can not be used on paraffin embedded tissue sections since the antigen is denatured during embedding and paraffin removal.

Immunocytochemistry:
0.5-10 μg/mL of a previous lot was used on 4% paraformaldehyde, acetic acid or ethanol fixed cultured cells.

ELISA:
A previous lot was used on purified galactocerebrosides.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal in buffer containing 10 mM Potassium Phosphate, 150 mM NaCl, pH 7.4 containing 0.09% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Neonatal cortex

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Matsushita et al.
Neuroscience, 136(1), 115-121 (2005-09-27)
The successive stages of development from oligodendrocyte progenitor to mature oligodendrocyte have been investigated in detail by using stage-specific antibodies. However, no cell lines are available that show stepwise differentiation from oligodendrocyte progenitors to mature oligodendrocytes. Here we show the
Oligodendrocyte dysfunction after induction of experimental anterior optic nerve ischemia.
Goldenberg-Cohen, N; Guo, Y; Margolis, F; Cohen, Y; Miller, NR; Bernstein, SL
Investigative Ophthalmology & Visual Science null
HIV binding, penetration, and primary infection in human cervicovaginal tissue.
Diane Maher, Xiaoyun Wu, Timothy Schacker, Julie Horbul, Peter Southern
Proceedings of the National Academy of Sciences of the USA null
Blockade of attachment and fusion receptors inhibits HIV-1 infection of human cervical tissue.
Hu, Q; Frank, I; Williams, V; Santos, JJ; Watts, P; Griffin, GE; Moore, JP; Pope, M; Shattock, RJ
The Journal of Experimental Medicine null
Adaptive HIV-specific B cell-derived humoral immune defenses of the intestinal mucosa in children exposed to HIV via breast-feeding.
Moussa, S; Jenabian, MA; Gody, JC; Leal, J; Gresenguet, G; Le Faou, A; Belec, L
Testing null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service