Skip to Content
Merck
All Photos(1)

Key Documents

O7139

Sigma-Aldrich

Monoclonal Anti-Oligodendrocyte Marker O4 antibody produced in mouse

clone O4, purified immunoglobulin, lyophilized powder

Synonym(s):

Oligodendrocyte Marker O4 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

O4, monoclonal

form

lyophilized powder

species reactivity

mouse, rat, chicken, human

technique(s)

flow cytometry: 0.25 μg/106 cells (rate differentiated cortical stem cells)
immunocytochemistry: 1-10 μg/mL

isotype

IgM

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OMG(4974)

General description

Monoclonal anti-oligodendrocyte marker O4 was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with white matter of the corpus callosum from bovine brain. Oligodendrocytes are myelinating cells in the central nervous system (CNS) that form the myelin sheath of axons to support rapid nerve conduction.

Immunogen

white matter of corpus callosum from bovine brain.

Application

Monoclonal anti-oligodendrocyte marker O4 antibody produced in mouse has been used in immunocytochemistry and immunofluorescence.

Biochem/physiol Actions

Oligodendrocyte marker O4 functions as a development marker of oligodendrocytes. The monoclonal anti-oligodendrocyte marker O4 antibody has been used to describe a biologically distinct stage of the oligodendroglial lineage in-vitro.†

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuroglial Cells in Long-Term Primary Cultures from the Gilthead Seabream (Sparus Aurata L.): New Functional In Vitro Model from Bony Fish Brain
Centoducati G, et al.
Italian Journal of Animal Science, 12 (2013)
Oligodendrocytes: biology and pathology
Bradl M, et al.
Acta Neuropathologica, 119, 37-53 (2010)
M2 microglia promotes neurogenesis and oligodendrogenesis from neural stem/progenitor cells via the PPAR? signaling pathway
Jichao Yuan
Oncotarget (2017)
Shuhui Yang et al.
ACS omega, 4(19), 18136-18146 (2019-11-14)
Neural stem cells (NSCs) play an important role in neural tissue engineering because of their capacity of self-renewal and differentiation to multiple cell lineages. The in vitro conventional neurosphere culture protocol has some limitations such as limited nutrition and oxygen
Xiao-Long Guo et al.
Communications biology, 2, 59-59 (2019-02-19)
Bacterial pore-forming toxin aerolysin-like proteins (ALPs) are widely distributed in animals and plants. However, functional studies on these ALPs remain in their infancy. βγ-CAT is the first example of a secreted pore-forming protein that functions to modulate the endolysosome pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service