CX2286
Cyclohexane
OmniSolv®
Synonym(s):
Hexahydrobenzene
About This Item
LC/MS: suitable
gas chromatography (GC): suitable
77 mmHg ( 20 °C)
Recommended Products
Agency
suitable for EPA 1613
Quality Level
vapor density
2.9 (vs air)
vapor pressure
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)
product line
OmniSolv®
Assay
≥99% (GC)
form
liquid
autoignition temp.
500 °F
expl. lim.
9 %
technique(s)
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
impurities
≤0.01% Water (H2O)
≤5 ng/L Capillary ECD responsive substances (as PCNB)
evapn. residue
≤1 ppm
color
APHA: ≤10
refractive index
n20/D 1.426 (lit.)
bp
80.7 °C (lit.)
mp
4-7 °C (lit.)
transition temp
flash point -18 °C
solubility
0.05 g/L
density
0.779 g/mL at 25 °C (lit.)
UV cutoff
≤200 nm
UV absorption
λ: 200 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.50
λ: 220 nm Amax: ≤0.20
λ: 230 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.05
λ: 250 nm Amax: ≤0.01
λ: 260 nm Amax: ≤0.01
λ: 270 nm Amax: ≤0.005
shipped in
ambient
storage temp.
room temp
SMILES string
C1CCCCC1
InChI
1S/C6H12/c1-2-4-6-5-3-1/h1-6H2
InChI key
XDTMQSROBMDMFD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Preparation Note
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service