Skip to Content
Merck
All Photos(1)

Key Documents

AX0142

Supelco

Acetonitrile

Acetonitrile

≥99.9% (GC), gradient grade, suitable for LC/MS, gas chromatography (GC), HPLC, OmniSolv®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12352117
EC Index Number:
200-835-2
Assay:
≥99.9% (GC)
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Acetonitrile, gradient grade OmniSolv®

vapor density

1.41 (vs air)

Quality Level

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water (H2O)
≤0.16 μeq/g Titrable base
≤20 ppm Propionitrile
≤8.0 μeq/g Titrable acid

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

UV cutoff

≤189 nm

UV absorption

λ: 189 nm Amax: ≤1.00
λ: 195 nm Amax: ≤0.15
λ: 210 nm Amax: ≤0.03
λ: 230 nm Amax: ≤0.005
λ: 250 nm Amax: ≤0.005
λ: 270 nm Amax: ≤0.005

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile is a water-soluble polar organic solvent with exceptional solubilizing power, unique chromatographic properties, and ideal spectroscopic characteristics. [1]

Application

Acetonitrile is used as a solvent:
  • To prepare mobile phase for LC/MS analysis of bioactive components in Graviola leaf extracts. [2]
  • In Reversed-phase HPLC. [3]
  • A solvent in chemical synthesis
  • A solvent for sample preparation, including extraction, dilution, and dissolution of compounds prior to analysis.

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)300ppt maxFormClear liquidGradient at 210 nm0.002AU maxGradient at 254 nm0.0003AU maxIdentity (IR-spectrum)ConformsPropionitrile20ppm maxRefractive index (at 20°C)1.343-1.345Titrable acid8.0µeq/g maxTitrable base0.16µeq/g maxUV Abs. at 189 nm1.00AU maxUV Abs. at 195 nm0.15AU maxUV Abs. at 210 nm0.03AU maxUV Abs. at 230 nm0.005AU maxUV Abs. at 250 nm0.005AU maxUV Abs. at 270 nm0.005AU maxUV Cut-off189nm maxWater (H2O)0.02% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioefficacy of Graviola leaf extracts in scavenging free radicals and upregulating antioxidant genes
Yu-Ra S, et al.
Food & Function, 7, 861-871 (2016)
A photoregulated ligand for the nuclear import receptor karyopherin alpha
Seung Bum P, et al.
Bioorganic & Medicinal Chemistry, 9, 3215-3223 (2001)
Comparison of the sensitivity of evaporative universal detectors and LC/MS in the HILIC and the reversed-phase HPLC modes
Clifford M R, et al.
Journal of chromatography, 877, 4133-4139 (2009)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service