Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
WX0004
Water
suitable for HPLC, OmniSolv®, meets USP/NF
About This Item
Recommended Products
Product Name
Water, gradient grade, Meets Reagent Specifications for testing USP/NF monographs OmniSolv®
grade
HPLC grade
Quality Level
Agency
meets USP/NF
vapor pressure
23 hPa ( 20 °C)
product line
OmniSolv®
form
liquid
shelf life
12 mo. (from manufacture date)
technique(s)
HPLC: suitable
impurities
≤5 ppb Total purgeable organics
evapn. residue
≤1 ppm
color
APHA: ≤10
refractive index
n20/D 1.34 (lit.)
bp
100 °C (lit.)
mp
0 °C (lit.)
density
1.000 g/mL at 3.98 °C (lit.)
UV absorption
λ: 190 nm Amax: ≤0.010
λ: 200 nm Amax: ≤0.010
λ: 210 nm Amax: ≤0.010
λ: 220 nm Amax: ≤0.010
λ: 230 nm Amax: ≤0.010
λ: 240 nm Amax: ≤0.010
λ: 250 nm Amax: ≤0.005
λ: 280 nm Amax: ≤0.005
application(s)
general analytical
shipped in
ambient
storage temp.
room temp
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Expiration Dated
Preparation Note
Analysis Note
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service