Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.06035
Methanol
≥99.9% (GC), suitable for LC/MS, LiChrosolv®
Synonym(s):
Methyl alcohol
About This Item
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Recommended Products
Product Name
Methanol, hypergrade for LC-MS LiChrosolv®
Agency
suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
Quality Level
reg. compliance
suitable for FDA C-010.02
vapor density
1.11 (vs air)
vapor pressure
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
product line
LiChrosolv®
Assay
≥99.9% (GC)
form
liquid
autoignition temp.
725 °F
potency
5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)
expl. lim.
36 %
technique(s)
LC/MS: suitable
impurities
≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water
≤10 ppb Al (Aluminium)
≤10 ppb Ca (Calcium)
≤10 ppb Fe (Iron)
≤10 ppb Mg (Magnesium)
≤100 ppb Na (Sodium)
≤5 ppb Any further metal (ICP-MS)
≤5 ppb K (Potassium)
evapn. residue
≤1.0 mg/L
SMILES string
CO
transmittance
210 nm, ≥35%
220 nm, ≥60%
230 nm, ≥75%
260 nm, ≥98%
refractive index
n20/D 1.329 (lit.)
kinematic viscosity
0.54-0.59 cSt(20 °C)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
transition temp
flash point 9.7 °C
density
0.791 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-30°C
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- The composition of chemicals and anti-osteogenic properties in the volatile extracts from Homalomena gigantea rhizome: This study highlights methanol′s role in the extraction and analysis of volatile organic compounds from medicinal plants, providing insights valuable for researchers in pharmacology and natural product chemistry (Thanh et al., 2024).
- A homochiral tartrate-bridged dinuclear chromium(III) complex anion with a resonance-assisted hydrogen bond for proton conduction: Methanol is used as a solvent in the synthesis of complex inorganic compounds, which are essential in developing new materials for energy applications, such as proton exchange membranes (Dunatov et al., 2024).
Preparation Note
Analysis Note
Identity (IR): conforms
Evaporation residue: ≤ 1.0 mg/l
Water: ≤ 0.01 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Al (Aluminium): ≤ 10 ppb
Ca (Calcium): ≤ 10 ppb
Fe (Iron): ≤ 10 ppb
K (Potassium): ≤ 5 ppb
Mg (Magnesium): ≤ 10 ppb
Na (Sodium): ≤ 100 ppb
Any further metal (ICP-MS): ≤ 5 ppb
Gradient grade (at 220 nm): ≤ 2.0 mAU
Gradient grade (at 235 nm): ≤ 1.0 mAU
Fluorescence (as quinine at 254 nm): ≤ 1.0 ppb
Fluorescence (as quinine at 365 nm): ≤ 0.5 ppb
Transmission (at 210 nm): ≥ 35 %
Transmission (at 220 nm): ≥ 60 %
Transmission (at 230 nm): ≥ 75 %
Transmission (from 260 nm): ≥ 98 %
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 2 ppb
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI negative): ≤ 20 ppb
Filtered by 0.2 µm filter.
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service