Skip to Content
Merck
All Photos(2)

Documents

1.09137

Supelco

Sodium hydroxide solution

c(NaOH) = 1 mol/l (1 N), reag. Ph. Eur., reag. USP, ready-to-use volumetric solution for titration, Titriplex®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

product name

Sodium hydroxide solution, c(NaOH) = 1 mol/l (1 N), Titripur®, reag. Ph. Eur., reag. USP

Agency

reag. Ph. Eur.
reag. USP

Quality Level

vapor pressure

3 mmHg ( 37 °C)

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Acid-base reactions

packaging

Titripac® of

concentration

1 M

technique(s)

titration: suitable

pH

13.7 (20 °C in H2O)

density

1.04 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Ozone-Activated Lignocellulose Films Blended with Chitosan for Edible Film Production.: This research involves the use of Sodium hydroxide solution in the preparation and activation of lignocellulose, which is then blended with chitosan to produce edible films, demonstrating a novel application in food packaging (Ebrahimi et al., 2024).

  • Tilapia Head Gelatins to Stabilize Fish Oil Emulsions and the Effect of Extraction Methods.: The study explores the use of Sodium hydroxide solution in the extraction process of gelatins from tilapia heads, which are then used to stabilize fish oil emulsions, highlighting an innovative use in food science (Li et al., 2024).

  • Study of Cellulose Dissolution in ZnO/NaOH/Water Solvent Solution and Its Temperature-Dependent Effect Using Molecular Dynamics Simulation.: This article investigates the dissolution of cellulose in a ZnO/NaOH/water solvent system, using molecular dynamics simulation to understand the temperature-dependent effects, showcasing the application of Sodium hydroxide solution in material science (Bourassi et al., 2024).

  • Potential for Recycling Metakaolin/Slag-Based Geopolymer Concrete of Various Strength Levels in Freeze-Thaw Conditions.: Sodium hydroxide solution is employed in the preparation of geopolymer concrete, which is analyzed for its recycling potential and durability under freeze-thaw conditions, emphasizing its application in sustainable construction materials (Liu et al., 2024).


Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Titripac® packaging - all SKU-pack size numbers ending "4000, 4003, 9010, 9013" - more information on www.sigmaaldrich.com/Titripac

Analysis Note

Form liquid
Amount-of-substance concentration 0.995 - 1.005 mol/L
Measurement uncertainty ± 0.003 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 84 potassium hydrogen phthalate) by means of volumetric standard potassium hydrogen phthalate (article number 1.02400), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service