Skip to Content
Merck
All Photos(2)

Documents

1.09959

Supelco

Sodium hydroxide solution

c(NaOH) = 0.1 mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
MDL number:
UNSPSC Code:
12352305
NACRES:
NA.21

product name

Sodium hydroxide solution, for 1000 ml, c(NaOH) = 0.1 mol/l (0.1 N) Titrisol®

vapor pressure

3 mmHg ( 37 °C)

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

13.8 (20 °C in H2O)

density

1.09 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[OH-].[Na+]

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Effective Management of Acute Oral Chemical Burns After NaOH Ingestion: A Case Report.: This case report describes the medical management of acute oral burns caused by ingestion of sodium hydroxide solution, providing critical insights into emergency response and treatment protocols in toxicology and emergency medicine (Li LB et al., 2024).

  • Mechanical Framework for Geopolymer Gels Construction: An Optimized LSTM Technique to Predict Compressive Strength of Fly Ash-Based Geopolymer Gels Concrete.: This study utilizes sodium hydroxide solution in the optimization of fly ash-based geopolymer gels, employing advanced machine learning techniques to predict material properties, which is pivotal in construction and materials engineering (Shi X et al., 2024).

  • Boosting-based ensemble machine learning models for predicting unconfined compressive strength of geopolymer stabilized clayey soil.: Research highlights the application of sodium hydroxide solution in soil stabilization processes. It focuses on using ensemble machine learning models to predict the strength of geopolymer-stabilized soils, contributing to geotechnical engineering advancements (Abdullah GMS et al., 2024).

  • Production of chitosan from shrimp shell using ultrasound followed by subcritical water hydrolysis.: This article discusses the use of sodium hydroxide solution in the extraction and purification process of chitosan from shrimp shells, highlighting its importance in biopolymer production and its potential applications in food technology and materials science (Matouri M et al., 2024).

  • A mesoionic carbene stabilized nickel(II) hydroxide complex: a facile precursor for C-H activation chemistry.: The study explores the synthesis of a nickel(II) hydroxide complex using sodium hydroxide solution, providing a new pathway for C-H activation in chemical synthesis, which is crucial for the development of catalytic processes and pharmaceutical manufacturing (Pavun A et al., 2024).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service