Skip to Content
Merck
All Photos(2)

Key Documents

1.06404

Supelco

Sodium chloride

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-598-3
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
358 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

1.3 hPa ( 865 °C)

product line

EMSURE®

Assay

≥99.5% (silver nitrate titration)
99-100.5% dry basis (silver nitrate titration)

form

solid

potency

3000 mg/kg LD50, oral (Rat)
>10000 mg/kg LD50, skin (Rabbit)

impurities

≤0.005% Calcium, Magnesium and R2O3-precipitate
≤0.005% Insoluble matter

loss

≤ 0.5% loss on drying, 105 °C, 2 h

pH

4.5-7.0 (20 °C, 100 g/L in H2O)

bp

1461 °C/1013 hPa

mp

801 °C (lit.)

solubility

358 g/L

bulk density

1140 kg/m3

anion traces

bromide (Br-): ≤0.005%
chlorate, nitrate (as NO3-): ≤0.003%
hexacyanoferrate(II) ([Fe(CN)6]4-): ≤0.0001%
iodide (I-): ≤0.001%passes test
nitrite (NO2-): passes test
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.001%

cation traces

As: ≤0.00004%
Ba: ≤0.001%, passes test
Ca: ≤0.002%
Cu: ≤0.0002%
Fe: ≤0.0001%
K: ≤0.005%
Mg: ≤0.001%
Mg: ≤0.0100% (alkaline-earth metals (as Ca))
N: ≤0.0005%
heavy metals: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Development of Low Sodium Table Butter via Partial Replacement of Sodium Chloride with Potassium Chloride.: This study explores reducing sodium content in table butter by substituting sodium chloride with potassium chloride. The findings are significant for chemists focused on food chemistry and the development of healthier food products with acceptable sensory characteristics (Biointerface Research in Applied Chemistry, 2020).

Analysis Note

Assay (argentometric): ≥ 99.5 %
Assay (argentometric; calculated on dried substance): 99.0 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Acidity or alkalinity: passes test
pH-value (5 %; water): 5.0 - 8.0
Insoluble matter: ≤ 0.005 %
Bromide (Br): ≤ 0.005 %
Chlorate and Nitrate (as NO₃): ≤ 0.003 %
Hexacyanoferrate II: ≤ 0.0001 %
Ferrocyanides: passes test
Iodide (I):
≤ 0.001 %
: passes test
Nitrite (NO₂): passes test
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
Heavy metals (ACS): ≤ 0.0005 %
As (Arsenic): ≤ 0.00004 %
Ba (Barium):
passes test
: ≤ 0.001 %
Ca (Calcium): ≤ 0.002 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0001 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.001 %
Calcium, Magnesium and R₂O₃-precipitate: ≤ 0.005 %
Magnesium and alkaline-earth metals (as Ca): ≤ 0.0100 %
Loss on drying (105 °C, 2 h): ≤ 0.5 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the packaging information for Item 1064049025?

    1 answer
    1. The packaging dimensions for Item 1064049025 are as follows:

      • Length: 37.00 cm
      • Width: 27.50 cm
      • Height: 31.00 cm

      Helpful?

  4. How to find SKU number for the COA? I know the catalogue number and the lot number, however what is the SKU number. The relevant product is sodium chloride, catalogue number: 1.06404.0500 and Lot: K49980304851. Thank you for your assistance.  

    1 answer
    1. The SKU number for this product is 1.06404.0500 the "-.0500" portion of this product number is the pack size which is 500 G. When retrieving the certificate of analysis for this product, customers will use lot K49980304. The last three digits "-851" of this lot is the packing code and should be omitted in order to obtain the CofA document from the website.

      Please navigate to this website to retrieve the COA:
      https://www.emdmillipore.com/product/Sodium-chloride,MDA_CHEM-106404#anchor_COA

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service