Skip to Content
Merck
All Photos(1)

Key Documents

1.05465

Millipore

MacCONKEY agar

for the isolation of Salmonella, Shigella and coliform bacteria (According harm. EP/USP/JP)

Synonym(s):

MACCONKEY agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Agency

according to EP
according to JP
according to USP

Quality Level

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

6.9-7.3 (25 °C, 50 g/L in H2O, after autoclaving)

solubility

50 g/L

bulk density

720 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Salmonella spp.
Shigella spp.
coliforms

General description

for the isolation of Salmonella, Shigella and coliform bacteria (According harm. EP/USP/JP)

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service