Skip to Content
Merck
All Photos(1)

Key Documents

70144

Millipore

MacConkey Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

bile salts, 5.0 g/L
lactose, 10.0 g/L
neutral red, 0.075 g/L
peptone, 20.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.2-7.6 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for Proteus spp.
selective and differential for Salmonella spp.
selective and differential for coliforms
selective and differential for enterobacteriaceae

Application

A presumptive medium for the detection of the coliform group in water and milk.

Preparation Note

Add 40 g to 1 litre of distilled water. Mix well and distribute into containers fitted with Durham tubes. Sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Recommendations of the Public Health Laboratory Service Water Subcommittee

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

EFFECT of anaerobic spore-bearing organisms on the validity of the presumptive coliform test as used in the bacteriological examination of water.
The Journal of hygiene, 51(2), 268-277 (1953-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service