Skip to Content
Merck
All Photos(1)

Documents

1.04316

Millipore

Buffered Peptone Water

NutriSelect® Prime, ISO 6579, ISO 6887, ISO 21528, ISO 22964, FDA BAM, EP, powder, Suitable for use in Halal and Kosher certified processes, pkg of 500 g

Synonym(s):

NutriSelect® prime Buffered Peptone Water halal acc. ISO 6579, ISO 6887, ISO 21528, ISO 22964, FDA-BAM and EP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP
ISO 21528
ISO 22964
ISO 6579
ISO 6887

Quality Level

reg. compliance

FDA BAM

form

powder

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.0 ± 0.2 (25.0 °C)

application(s)

food and beverages

storage temp.

15-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

General description

NutriSelect® prime Buffered Peptone Water halal is rich in nutrients and produces high resuscitation rates for sublethal injured bacteria and intense growth.
The phosphate buffer system maintains the pH during the pre-enrichment period and prevents bacterial damage caused by changes in the pH of the medium.
Peptone acts as a source of carbon, nitrogen, vitamins, and minerals whilst sodium chloride maintains the osmotic balance.

Application

NutriSelect® prime Buffered Peptone Water halal is used for the preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter spp., and for the initial suspension and dilution of samples from food and animal feed, water, and other materials

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service