Skip to Content
Merck
All Photos(1)

Key Documents

1.04327

Millipore

NutriSelect® prime YGC/OGY Agar (base) halal (Yeast extract Glucose Agar base) without antibiotics, acc. ISO 6611 I IDF 94

Suitable for use in Halal and Kosher certified processes, NutriSelect® Prime

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 6611 I

Quality Level

form

powder

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

6.6 ± 0.2 (25.0 °C)

application(s)

food and beverages

storage temp.

15-25°C

suitability

selective for molds

General description

NutriSelect® prime Yeast Extract Glucose Chloramphenicol (YGC/OGY) Agar halal is a nutrient rich selective agar. The media composition contains antimicrobial agent like chloramphenicol (YGC Agar) or oxytetracycline to inhibit accompanying bacterial growth.
It is fully autoclavable along with chloramphenicol, unlike other microbial culture media.
The medium contains yeast extract which provides the vitamin source, B-complex. Glucose supplies energy for the growth of yeasts and molds and agar is the solidifying agent.

Application

NutriSelect® prime Yeast Extract Glucose Chloramphenicol (YGC/OGY) Agar halal is used for enumeration of yeasts and molds in milk, dairy products and other food and feeding stuff as well as from environmental samples from food production and food handling.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service