Skip to Content
Merck
All Photos(1)

Key Documents

Z-004

Supelco

Zaleplon solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15N5O
CAS Number:
Molecular Weight:
305.33
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CCN(C(C)=O)c1cccc(c1)-c2ccnc3c(cnn23)C#N

InChI

1S/C17H15N5O/c1-3-21(12(2)23)15-6-4-5-13(9-15)16-7-8-19-17-14(10-18)11-20-22(16)17/h4-9,11H,3H2,1-2H3

InChI key

HUNXMJYCHXQEGX-UHFFFAOYSA-N

Gene Information

General description

Zaleplon is a nonbenzodiazepine “Z-drug” prescribed under the trade names Sonata® or Starnoc for the treatment of insomnia. Zaleplon is often abused illicitly for its mind-altering effects. This Certified Spiking Solution® is applicable to use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Sonata is a registered trademark of King Pharmaceuticals Research and Development, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Shu Huang et al.
Chang Gung medical journal, 34(1), 50-56 (2011-03-12)
Benzodiazepines cause a high proportion of adverse effects while non-benzodiazepine compounds have demonstrated high efficacy and less adverse effects in patients with insomnia. The objective of this study was to compare the effectiveness and safety of non-BZ zaleplon and zolpidem
A case of zaleplon-induced amnestic sleep-related eating disorder.
Shanna M Molina et al.
The Journal of clinical psychiatry, 71(2), 210-211 (2010-03-03)
Ane Birgitte Telén Andersen et al.
Pharmacoepidemiology and drug safety, 20(4), 378-385 (2011-01-25)
To estimate the degree of long-term use of zopiclone, zolpidem and zaleplon among Danes aged 65 and older and the association with sociodemographic factors and use of other drugs. Danish register-based study of 5000 men and 5000 women aged 65
Insomnia: evidence-based approaches to assessment and management.
Kevin Morgan et al.
Clinical medicine (London, England), 11(3), 278-281 (2011-09-10)
Jasna Jablan et al.
Journal of pharmaceutical and biomedical analysis, 71, 35-44 (2012-08-18)
The aim of this work was to investigate the potential synergistic effect of water-soluble polymers (hypromellose, HPMC and polyvinylpyrrolidone, PVP) on zaleplon (ZAL) complexation with parent β-cyclodextrin (βCD) and its randomly methylated derivative (RAMEB) in solution and in solid state.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service