Skip to Content
Merck
All Photos(2)

Key Documents

W103

Sigma-Aldrich

WIN 51708 hydrate

solid

Synonym(s):

17-β-Hydroxy-17-α-ethynyl-5-α-androstano[3,2-β]pyrimido[1,2-α]benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H33N3O · xH2O
CAS Number:
Molecular Weight:
439.59 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

yellow

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

O.C[C@]12Cc3cn4c(nc3C[C@@H]1CC[C@@H]5[C@@H]2CC[C@@]6(C)[C@H]5CC[C@@]6(O)C#C)nc7ccccc47

InChI

1S/C29H33N3O.H2O/c1-4-29(33)14-12-22-20-10-9-19-15-24-18(16-27(19,2)21(20)11-13-28(22,29)3)17-32-25-8-6-5-7-23(25)30-26(32)31-24;/h1,5-8,17,19-22,33H,9-16H2,2-3H3;1H2/t19-,20+,21-,22-,27-,28-,29-;/m0./s1

InChI key

RGVGINOANKXLLR-GCGDSGHKSA-N

Gene Information

Biochem/physiol Actions

WIN 51708 hydrate is a non-peptide NK1 tachykinin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Appell et al.
Molecular pharmacology, 41(4), 772-778 (1992-04-01)
125I-Bolton-Hunter-substance P (125I-BH-SP) binding properties of three novel classes of neurokinin-1 (NK-1) receptor antagonists were investigated in tissues derived from humans, guinea pigs, and rats. 125I-BH-SP was shown to bind to a single class of binding sites, with similar dissociation
B R Venepalli et al.
Journal of medicinal chemistry, 35(2), 374-378 (1992-01-24)
Several heterosteroids containing a dihydroethisterone skeleton were prepared and shown to displace substance P in a receptor binding assay. Further biochemical (kinetic and Scatchard analyses) and pharmacological evaluation (substance P-induced plasma extravasation and salivation in the rat) of a representative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service