Skip to Content
Merck
All Photos(1)

Documents

ERT-107S

Supelco

2,4,6-Trinitrotoluene solution

10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®

Synonym(s):

TNT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O6
CAS Number:
Molecular Weight:
227.13
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 5 mL

manufacturer/tradename

Cerilliant®

concentration

10 mg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1[N+]([O-])=O

InChI

1S/C7H5N3O6/c1-4-6(9(13)14)2-5(8(11)12)3-7(4)10(15)16/h2-3H,1H3

InChI key

SPSSULHKWOKEEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Trinitrotoluene (TNT) solution is a certified reference material, suitable for the in vitro identification, calibration and quantification of the analyte(s) in analytical and R & D applications. Not suitable for human or animal consumption.

Application

2,4,6-Trinitrotoluene solution has been used as a reference material to investigate its detection and identification post-deposition on a surface using quantum cascade lasers (QCLs) in combination with photoacoustic spectroscopy (PAS).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoacoustic spectroscopy for chemical detection
Chemical, Biochemical and Medical Aspects, 8358, 83581I-83581I (2012)
Craig A McFarland et al.
Ecotoxicology (London, England), 21(8), 2372-2390 (2012-09-15)
Evaluation of multiple-stressor effects stemming from habitat degradation, climate change, and exposure to chemical contaminants is crucial for addressing challenges to ecological and environmental health. To assess the effects of multiple stressors in an understudied taxon, the western fence lizard
Tommaso Rossi et al.
Investigative ophthalmology & visual science, 53(13), 8057-8066 (2012-11-01)
Primary blast injury (PBI) mostly affects air-filled organs, although it is sporadically reported in fluid-filled organs, including the eye. The purpose of the present paper is to explain orbit blast injury mechanisms through finite element modeling (FEM). FEM meshes of
Liangdong Zou et al.
Applied biochemistry and biotechnology, 168(7), 1976-1988 (2012-10-19)
2,4,6-Trinitrotoluene (TNT), an extensively used and versatile explosive, is harmful in soil and water. In the present study, four bacterial strains capable of degrading TNT have been isolated from contaminated sites and named as Thu-A, Thu-B, Thu-C, and Thu-Z. Thu-Z
Ronit Freeman et al.
Advanced materials (Deerfield Beach, Fla.), 24(48), 6416-6421 (2012-09-26)
Chemically modified CdSe/ZnS quantum dots (QDs) are used as fluorescent probes for the analysis of explosives, and specifically, the detection of trinitrotoluene (TNT) or trinitrotriazine (RDX). The QDs are functionalized with electron-donating ligands that bind nitro-containing explosives, exhibiting electron-acceptor properties

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service