Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,4,6-Trinitrotoluene solution
Empirical Formula (Hill Notation):
C
7
H
5
N
3
O
6
CAS Number:
118-96-7
Molecular Weight:
227.13
Products
Brand
Supelco (2)
Formula Weight
Manufacturer
Format
Application
Package Size
Reference Material Type
Special Grade
certified reference material (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
ERT-022S
2,4,6-Trinitrotoluene solution,
1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant
®
Expand
View Pricing
ERT-107S
2,4,6-Trinitrotoluene solution,
10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant
®
Expand
View Pricing