Skip to Content
Merck
All Photos(1)

Documents

P-037

Supelco

Pentaerythritol tetranitrate solution

1000 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Pentaerythritol tetranitrate, PETN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N4O12
CAS Number:
Molecular Weight:
316.14
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1000 μg/mL in acetonitrile

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

O=[N+]([O-])OCC(CO[N+]([O-])=O)(CO[N+]([O-])=O)CO[N+]([O-])=O

InChI

1S/C5H8N4O12/c10-6(11)18-1-5(2-19-7(12)13,3-20-8(14)15)4-21-9(16)17/h1-4H2

InChI key

TZRXHJWUDPFEEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Options of NO substitution: proven and future methods. A pharmaceutical classic on the upswing].
MMW Fortschritte der Medizin, 153(24-25), 50-51 (2011-07-02)
Li Zhuang et al.
Chemosphere, 89(7), 810-816 (2012-06-01)
This study examined the role of denitrifying and sulfate-reducing bacteria in biodegradation of pentaerythritol tetranitrate (PETN). Microbial inocula were obtained from a PETN-contaminated soil. PETN degradation was evaluated using nitrate and/or sulfate as electron acceptors and acetate as a carbon
Swenja Schuhmacher et al.
Hypertension (Dallas, Tex. : 1979), 55(4), 897-904 (2010-02-17)
The organic nitrate pentaerythritol tetranitrate is devoid of nitrate tolerance, which has been attributed to the induction of the antioxidant enzyme heme oxygenase (HO)-1. With the present study, we tested whether chronic treatment with pentaerythritol tetranitrate can improve angiotensin II-induced
Monica Lisi et al.
Clinical research in cardiology : official journal of the German Cardiac Society, 101(6), 453-459 (2012-02-03)
Ischemic and pharmacologic preconditioning have great clinical potential, but it remains unclear whether their effects can be maintained over time during repeated exposure.We have previously demonstrated that the acute protective effect of nitroglycerin (GTN) is attenuated during repeated daily administration.
A Edtbauer et al.
The Journal of chemical physics, 132(13), 134305-134305 (2010-04-15)
Gas phase dissociative electron attachment (DEA) measurements to pentaerythritol tetranitrate (PETN) are performed in a crossed electron-molecular beam experiment at high-energy resolution and high sensitivity. DEA is operative at very low energies close to approximately 0 eV showing unique features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service